Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, 8 mm Hole, Aluminum, Silicone Transparent Blue/PTFE White, 45° Shore A, 1.3 mm Septa, Silver
Catalog # 97054-914
Supplier #:  95020-45S-T
Headspace Caps, MicroSolv
Catalog # 97054-914
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, 8 mm Hole, Aluminum, Silicone Transparent Blue/PTFE White, 45° Shore A, 1.3 mm Septa, Silver
Price
$309.17
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Silicone Transparent Blue/PTFE White, 45° Shore A, 1.3 mm Septa
  • Color:
    Silver
  • Description:
    Magnetic Screw Cap
  • Material:
    Aluminum
  • Size:
    18 mm Precision Threads, 8 mm Hole