Orders LinkContactUsLinkComponent
Screw Caps, MicroSolv™
PP, Red-Orange Silicone Rubber / TEF, 9-425 mm, Blue, Screw cap, solid
Catalog # 97052-482
 :  9502S-09-SR
Screw Caps, MicroSolv™
Catalog # 97052-482
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
PP, Red-Orange Silicone Rubber / TEF, 9-425 mm, Blue, Screw cap, solid
Price
$44.75
Log in for availability
Pack of 100
Product Details & Documents
Specifications
  • Cap Size:
    9-425 mm
  • Cap type:
    Screw cap, solid
  • Septum:
    Red-Orange Silicone Rubber / TEF
  • Material:
    PP
  • Cap color:
    Blue