Orders LinkContactUsLinkComponent
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
PP, Bonded Silicone/PTFE, low durometer, 9-425 mm, Blue, Screw cap, knurled
Catalog # 97051-422
Supplier #:  9502S-10C-B
Screw Caps, MicroSolv™
Catalog # 97051-422
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
PP, Bonded Silicone/PTFE, low durometer, 9-425 mm, Blue, Screw cap, knurled
Price
$50.58
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Specifications
  • Cap Size:
    9-425 mm
  • Cap type:
    Screw cap, knurled
  • Septum:
    Bonded Silicone/PTFE, low durometer
  • Material:
    PP
  • Cap color:
    Blue