Orders LinkContactUsLinkComponent
Solid Polyethylene Acid Cabinets, Justrite®
Solid Polyethylene Acid Cabinets, Justrite®
Acids underbench cabinet, 2 door, undercounter, 36× 2½ L bottles, Polyethylene
Catalog # 97048-666
Supplier:   Justrite
Supplier #:  24010
Solid Polyethylene Acid Cabinets, Justrite®
Catalog # 97048-666
Supplier:  Justrite
Solid Polyethylene Acid Cabinets, Justrite®
Solid Polyethylene Acid Cabinets, Justrite®
Acids underbench cabinet, 2 door, undercounter, 36× 2½ L bottles, Polyethylene
Price
$10,402.71
Log in for availability
Each
Product Details & Documents
These solid wall high density polyethylene cabinets have excellent chemical resistance to handle the most corrosive chemicals.
  • Sturdy, smooth construction in an attractive design
  • Suitable for highly aggressive liquids - no metal parts to corrode
  • Three-point door latch on larger units offer secure closure
  • Toe kick on undercounter model allows user to stand closer to counter - less leaning, better ergonomics
They are 100% poly welded with no metal parts, they include a external 2" NPT vent/exhaust connections and have padlockable hasps for security. White.
Documents
Specifications
  • Capacity:
    2 door, undercounter, 36× 2½ L bottles
  • Description:
    Acids underbench cabinet
  • Material:
    Polyethylene