Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER TECL DIN 19 267 PH25C-10.00 25ML
Catalog # 97042-534
Supplier:
SI Analytics
Expand image
BUFFER TECL DIN 19 267 PH25C-10.00 25ML
Catalog # 97042-534
Supplier:
SI Analytics
Supplier Number:
285138205