Orders LinkContactUsLinkComponent
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Pepsin/Hydrochloric Acid Solution, 1000 mL, Glass bottle, DURAN®
Catalog # 97042-510
Supplier:   SI Analytics
Supplier #:  285138538
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Catalog # 97042-510
Supplier:  SI Analytics
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Pepsin/Hydrochloric Acid Solution, 1000 mL, Glass bottle, DURAN®
Price
$246.05
Log in for availability
Each
Product Details & Documents
Suitable for cleaning combination electrodes and reference electrodes.
Documents
Specifications
  • Description:
    Pepsin/Hydrochloric Acid Solution
  • Size:
    1000 mL
  • Packaging:
    Glass bottle, DURAN®