Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLUTION ELECT LICL 0.1MOL/L ETOH 250ML
Catalog # 97042-454
Supplier:
SI Analytics
SDS
Expand image
SOLUTION ELECT LICL 0.1MOL/L ETOH 250ML
Catalog # 97042-454
Supplier:
SI Analytics
Supplier Number:
285138308
SDS
Related Information
Documentation
SDS
ghs_285138308.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation