$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER STD DIN 19 267 PH25C-7 250ML
97042-444
:
SI Analytics
BUFFER STD DIN 19 267 PH25C-7 250ML
97042-444
:
SI Analytics
:
285138032