Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER STD DIN 19 266 PH25C-6.87 250ML
Catalog # 97042-440
Supplier:
SI Analytics
Expand image
BUFFER STD DIN 19 266 PH25C-6.87 250ML
Catalog # 97042-440
Supplier:
SI Analytics
Supplier Number:
285138016