Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH METER PROLAB 1000 BNC
Catalog # 97042-336
Supplier:
SI Analytics
Expand image
PH METER PROLAB 1000 BNC
Catalog # 97042-336
Supplier:
SI Analytics
Supplier Number:
285204200
Related Information
Documentation
Manual
ProLab-1000 Digital-pH-Meter 1.1-MB
Documentation