$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
PH METER LAB860 BNC SET/ILA170
97042-322
:
SI Analytics
PH METER LAB860 BNC SET/ILA170
97042-322
:
SI Analytics
:
285204130