Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco

Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco

Storage Shelf Kit, Left-Side Interior, No
  97002-466
 :   Labconco
Supplier #:  5235001
Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco
  97002-466
 :  Labconco
Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco

Storage Shelf Kit, Left-Side Interior, No

 
$610.00
   
Each
Minimum order quantity is 1
 
  • Description:
    Storage Shelf Kit, Left-Side Interior
  • Rediship:
    No
Accessories

 

  •  
    Description
    Electrical
    For use with
     
     
  • 97002-432
    Diaphragm Vacuum Pump, 42 L/min displacement at 60Hz
    115 V, 60 Hz, 3.3 A (US plug)
    Protector and Precise Glove Box Drying Trains
       
    Each - $2,190.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 97002-434
    Diaphragm Vacuum Pump, 35 L/min displacement at 50Hz
    230 V, 50 Hz, 1.5 A (US plug)
    Protector and Precise Glove Box Drying Trains
       
    Each - $2,260.00

 

  •  
    Description
    For use with
     
     
  • 83009-292
    Insert, Ammonia Trap
    Drying Train for Precise Glove Boxes
       
    Each - $169.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 26671-492
    Vacuum Pump, 117 L/min. displacement at 60 Hz
    230 V, 50/60 Hz, 2.4 A (230 V reverse IEC plug)
    FreeZone Freeze Dryers (230V) 4.5L and smaller, CentriVap Concentrators, and Protector and Precise CA Glove Boxes
       
    Each - $4,340.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 26671-484
    Vacuum Pump, 117 L/min. displacement at 60 Hz
    115 V, 50/60 Hz, 4.6 A (NEMA 5-15P)
    FreeZone Freeze Dryers (115V) 4.5L and smaller, CentriVap Concentrators, and Protector and Precise CA Glove Boxes
       
    Each - $4,220.00

 

  •  
    Description
    Type
    Width
     
     
  • 46610-462
    Standard base cabinet (for precise glove boxes)
    2 doors
    30"
       
    Each - $2,190.00

 

  •  
    Description
    Glove size
    Glove color
     
     
  • 32948-205
    Butyl gloves
    9.75
    Black
       
    Pack of 2 - $880.00

 

  •  
    Description
    Glove size
    Glove color
     
     
  • 32948-019
    Neoprene gloves
    9.75
    Black
       
    Pair - $625.00

 

  •  
    Description
    For use with
     
     
  • 26675-884
    Insert, Solvent Trap
    Drying Train for Precise Glove Boxes
       
    Each - $172.00

 

  •  
    Description
    For use with
     
     
  • 26675-881
    Insert, Radiochemical
    Drying Train for Precise Glove Boxes
       
    Each - $198.00

 

  •  
    Description
    For use with
     
     
  • 26675-879
    Insert, Moisture Trap
    Drying Train for Precise Glove Boxes
       
    Each - $172.00

 

  •  
    Description
    For use with
     
     
  • 26675-878
    Insert, Acid Trap
    Drying Train for Precise Glove Boxes
       
    Each - $172.00