Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco
Oxygen and Moisture Removal Column, No
Catalog # 97002-436
Supplier: Labconco
: 5244200
Accessories for Precise™ Basic and Controlled Atmosphere Glove Boxes, Labconco
Catalog # 97002-436
Supplier: Labconco
Jump to:
Product Details & Documents
Specifications
- Description:Oxygen and Moisture Removal Column
- Rediship:No
Accessories
Product Information
- Catalog #ElectricalFor use withAvailabilityPrice
- Diaphragm Vacuum Pump, 42 L/min displacement at 60HzDiaphragm Vacuum Pump, 42 L/min displacement at 60Hz97002-432115 V, 60 Hz, 3.3 A (US plug)Protector and Precise Glove Box Drying TrainsLog in for availabilityEach - $2,190.00
Product Information
- Catalog #ElectricalFor use withAvailabilityPrice
- Diaphragm Vacuum Pump, 35 L/min displacement at 50HzDiaphragm Vacuum Pump, 35 L/min displacement at 50Hz97002-434230 V, 50 Hz, 1.5 A (US plug)Protector and Precise Glove Box Drying TrainsLog in for availabilityEach - $2,260.00
Product Information
- Catalog #For use withAvailabilityPrice
- Insert, Ammonia TrapInsert, Ammonia Trap83009-292Drying Train for Precise Glove BoxesLog in for availabilityEach - $169.00
Product Information
- Catalog #ElectricalFor use withAvailabilityPrice
- Vacuum Pump, 117 L/min. displacement at 60 HzVacuum Pump, 117 L/min. displacement at 60 Hz26671-492230 V, 50/60 Hz, 2.4 A (230 V reverse IEC plug)FreeZone Freeze Dryers (230V) 4.5L and smaller, CentriVap Concentrators, and Protector and Precise CA Glove BoxesLog in for availabilityEach - $4,340.00
Product Information
- Catalog #ElectricalFor use withAvailabilityPrice
- Vacuum Pump, 117 L/min. displacement at 60 HzVacuum Pump, 117 L/min. displacement at 60 Hz26671-484115 V, 50/60 Hz, 4.6 A (NEMA 5-15P)FreeZone Freeze Dryers (115V) 4.5L and smaller, CentriVap Concentrators, and Protector and Precise CA Glove BoxesLog in for availabilityEach - $4,220.00
Product Information
- Catalog #TypeWidthAvailabilityPrice
- Standard base cabinet (for precise glove boxes)Standard base cabinet (for precise glove boxes)46610-4622 doors30"Log in for availabilityEach - $2,190.00
Product Information
- Catalog #Glove sizeGlove colorAvailabilityPrice
- Butyl gloves
Product Information
- Catalog #Glove sizeGlove colorAvailabilityPrice
- Neoprene gloves
Product Information
- Catalog #For use withAvailabilityPrice
- Insert, Solvent TrapInsert, Solvent Trap26675-884Drying Train for Precise Glove BoxesLog in for availabilityEach - $172.00
Product Information
- Catalog #For use withAvailabilityPrice
- Insert, RadiochemicalInsert, Radiochemical26675-881Drying Train for Precise Glove BoxesLog in for availabilityEach - $198.00
Product Information
- Catalog #For use withAvailabilityPrice
- Insert, Moisture TrapInsert, Moisture Trap26675-879Drying Train for Precise Glove BoxesLog in for availabilityEach - $172.00
Product Information
- Catalog #For use withAvailabilityPrice
- Insert, Acid TrapInsert, Acid Trap26675-878Drying Train for Precise Glove BoxesLog in for availabilityEach - $172.00