Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
NMC Valve Wheel Lockouts

NMC Valve Wheel Lockouts

Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.
Catalog # 94022-908
Supplier #:  VL06R
NMC Valve Wheel Lockouts
Catalog # 94022-908
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts

Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.

Price
$126.17
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter

Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F

Specifications
  • Description:
    Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.