NMC Mini Lockout Center
Lockout Center Equipped
Catalog # 94017-526
Supplier: ACCUFORM MANUFACTURING, INC
: MLO1
Jump to:
Product Details & Documents
Made from .125 plexiglass and features 8 replaceable plastic hooks and a clear plastic pocket
Dimensions: 39.37cm x 35.56cm (15½" x 14") Available with or without lockout supplies
Specifications
- Description:Lockout Center Equipped