Orders LinkContactUsLinkComponent
Replacement Tips, Excelta Corp®
Replacement Tips, Excelta Corp®
Replacement tips for 759-RT, 3 star quality, Carbon fiber tips, 0.63 mm (0.025"), 0.012" (0.3 mm), 51° curved
Catalog # 94002-680
Replacement Tips, Excelta Corp®
Catalog # 94002-680
Supplier:  
Replacement Tips, Excelta Corp®
Replacement Tips, Excelta Corp®
Replacement tips for 759-RT, 3 star quality, Carbon fiber tips, 0.63 mm (0.025"), 0.012" (0.3 mm), 51° curved
Discontinued
Product Details & Documents
High-quality forcep tips.
  • High rigidity, excellent tensile and flexural strength.
These soft replacement tips are made of a Carbofib™ material which is a polymer based material with 30% carbon fibers added. The working temperature is 249 °F. Carbofib™ has very high rigidity, excellent tensile and flexural strength, and fatigue and creep resistance. The tips are conductive (102 ohms/sq). Each includes 4 screws and 2 tips.
Specifications
  • Description:
    Replacement tips for 759-RT, 3 star quality
  • Material:
    Carbon fiber tips
  • Curvature:
    51° curved
  • Tip width:
    0.012" (0.3 mm)
  • Tip length:
    0.63 mm (0.025")