Orders LinkContactUsLinkComponent
Replacement Tips, Excelta Corp®
Replacement tips for 159C-RT, 3 star quality, Carbon fiber tips, 1.38 mm (0.055"), 0.08" (0.2 mm), Straight
  94002-676
 :   Excelta
 :  159C-RTX
Replacement Tips, Excelta Corp®
  94002-676
 :  Excelta
Replacement Tips, Excelta Corp®
Replacement Tips, Excelta Corp®
Replacement tips for 159C-RT, 3 star quality, Carbon fiber tips, 1.38 mm (0.055"), 0.08" (0.2 mm), Straight
$11.76
Each
High-quality forcep tips.
  • High rigidity, excellent tensile and flexural strength.
These soft replacement tips are made of a Carbofib™ material which is a polymer based material with 30% carbon fibers added. The working temperature is 249 °F. Carbofib™ has very high rigidity, excellent tensile and flexural strength, and fatigue and creep resistance. The tips are conductive (102 ohms/sq). Each includes 4 screws and 2 tips.
  • Description:
    Replacement tips for 159C-RT, 3 star quality
  • Material:
    Carbon fiber tips
  • Curvature:
    Straight
  • Tip width:
    0.08" (0.2 mm)
  • Tip length:
    1.38 mm (0.055")