Order Entry
Puerto Rico
ContactUsLinkComponent
INCUBATOR REFRIG REACH-IN CO2 230V 708L
Catalog # 89174-560
Supplier:  Caron Products
undefined
INCUBATOR REFRIG REACH-IN CO2 230V 708L
Catalog # 89174-560
Supplier:  Caron Products
Supplier Number:  6026-3

Related Information