Orders LinkContactUsLinkComponent
Duramark® Lock Boxes
Duramark® Lock Boxes
Plastic lock box with over-door strap, 34.3×26×17.1 cm (13½×10¼× 6¾”)
Catalog # 89170-984
Supplier:   Therapak, LLC
Supplier #:  36506
Duramark® Lock Boxes
Catalog # 89170-984
Supplier:  Therapak, LLC
Duramark® Lock Boxes
Duramark® Lock Boxes
Plastic lock box with over-door strap, 34.3×26×17.1 cm (13½×10¼× 6¾”)
Price
$279.41
Log in for availability
Each
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Specifications
  • Description:
    Plastic lock box with over-door strap
  • Dimensions:
    34.3×26×17.1 cm (13½×10¼× 6¾”)
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Under-Door Strap for Metal or Plastic Box
    Under-Door Strap for Metal or Plastic Box
    89170-896
    Log in for availability
    Each - $40.90
Product Information
  • Catalog #
    Availability
    Price
  • Over-Door Strap for Metal or Plastic Box
    Over-Door Strap for Metal or Plastic Box
    89170-894
    Log in for availability
    Each - $35.21
Product Information
  • Catalog #
    Availability
    Price
  • Extra Long Over-Door Strap for Metal or Plastic Box
    Extra Long Over-Door Strap for Metal or Plastic Box
    10497-106
    Log in for availability
    Each - $118.05
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Key for Metal or Plastic Box
    Replacement Key for Metal or Plastic Box
    89170-898
    Log in for availability
    Each - $9.02