Corning® Bar Coded Storage Tubes, Corning
384× 2D barcoded storage tubes, round, without plug caps, Black, 110 µl, racked
Catalog # 89168-524
Supplier: Corning
: 8509
Jump to:
Product Details & Documents
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
- Maximum information, 14×14 dot 2D barcode
- Barcode stability, laser-etched, not an attached label
- Compound compatibility, polypropylene construction throughout, making it inert
Certificates
Search by Lot NumberSpecifications
- Volume:110 µl
- Description:384× 2D barcoded storage tubes, round, without plug caps, Black
- Packaging:racked
Accessories
Product Information
- Catalog #AvailabilityPrice
- 96 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 850096 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 850089168-514Log in for availabilityCase of 4,800 - $3,960.00
Product Information
- Catalog #AvailabilityPrice
- 96 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 850096 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 850089168-516Log in for availabilityCase of 4,800 - $4,610.00
Product Information
- Catalog #AvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 8500Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 850089168-518Log in for availabilityCase of 9,600 - $826.00
Product Information
- Catalog #AvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 8500Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 850089168-520Log in for availabilityCase of 4,800 - $1,160.00
Product Information
- Catalog #AvailabilityPrice
- Empty racks with lids for 96 screw capped storage tubes no. 8502, NaturalEmpty racks with lids for 96 screw capped storage tubes no. 8502, Natural89168-522Log in for availabilityCase of 10 - $114.00
Product Information
- Catalog #AvailabilityPrice
- 384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, Orange384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, Orange89168-530Log in for availabilityCase of 38,400 - $5,560.00
Product Information
- Catalog #AvailabilityPrice
- 384 Storage Tube Plug Cap,Orange, for no. 8509384 Storage Tube Plug Cap,Orange, for no. 850989168-528Log in for availabilityCase of 38,400 - $3,980.00
Product Information
- Catalog #AvailabilityPrice
- Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 8510Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 851089168-532Log in for availabilityCase of 20 - $286.00