Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
89168-396
Objectives for Motic AE2000 Inverted Compound Microscope, Motic Instruments
89168-396
- Description:CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8