ContactUsLinkComponent
CFDA SE Cell Prol. Kit
ViaFluor® SE Cell Proliferation Kits, Biotium
Catalog # 89138-974
Supplier:  Biotium
CAS Number:  
ViaFluor® SE Cell Proliferation Kits, Biotium
Catalog # 89138-974
Supplier:  Biotium
Supplier Number:  30050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ViaFluor® CFSE Cell Proliferation Kit, Ex/Em: 495/519 nm (Dye)
  • Size:
    90 assays
  • Cat. No.:
    89138-974
  • Supplier no.:
    30050
Specifications
About this item
Fluorescent dyes for tracking cell divisions by flow cytometry or tracing live cells by microscopy.
  • Non-toxic dyes covalently label cell cytoplasm
  • Track cell proliferation by dye dilution in flow cytometry
  • Long term imaging of cell morphology or tracking cells in co-culture by microscopy
  • Violet ViaFluor® 405 offers excellent performance at lower cost
  • Green ViaFluor® 488 SE is much less toxic than CFSE
ViaFluor® SE Cell Proliferation Kits use amine-reactive dyes to covalently label cells throughout the cell cytoplasm and intracellular compartments. Cell proliferation dyes are commonly used to monitor cell division by flow cytometry. The dyes also can be used to stably label cells to image cell morphology, or to track cell populations in mixed co-culture experiments. Labeling is covalent, so it withstands fixation and permeabilization for subsequent immunostaining.
The membrane permeable ViaFluor® SE compound is non-fluorescent until it enters viable cells, where it is hydrolyzed by cytoplasmic esterase enzymes to releases the fluorescent amine-reactive dye. The dyes then covalently react with amine groups on intracellular proteins, forming fluorescent conjugates that are retained in the cell, while excess unreacted dye is washed away. Immediately after staining, a single, bright fluorescent population will be detected by flow cytometry. With each cell division, daughter cells inherit roughly half of the fluorescent label, allowing the number of cell divisions that occur after labeling to be detected by the appearance of successively dimmer fluorescent peaks on a flow cytometry histogram compared to cells analyzed immediately after staining. Thus, cell proliferation dyes can be used to track multiple cell divisions of cells grown in culture or injected in vivo after labeling with the ViaFluor® SE dye.
ViaFluor® SE dyes also can be used for imaging cell morphology or identifying cells in co-culture by microscopy. Because the staining is non-toxic and well-retained, it can be used for imaging live cells over time.
Ordering information: Trial size kits include 1 vial of dye and 1 vial of anhydrous DMSO for dissolving the dye. Regular size kits include 10 dye vials and 1 vial of anhydrous DMSO.