Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

PYREX® Hydrometer Cylinders, Plain, Corning

300 ml, 39 mm, 381 mm
Catalog # 89091-314
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
 :  2962-300
PYREX® Hydrometer Cylinders, Plain, Corning
Catalog # 89091-314
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Hydrometer Cylinders, Plain, Corning
PYREX® Hydrometer Cylinders, Plain, Corning

300 ml, 39 mm, 381 mm

Price
$730.00
Log in for availability
Case of 12
Product Details & Documents

These PYREX® glas hydrometer cylinders have a large hexagonal base sealed to the cylinder body to increase stability. Breakage is minimized by the heavy wall construction.

  • Excellent chemical resistance
  • Good temperature resistance

Meets ASTM specifications D-287 and E-100.

Specifications
  • Ø:
    39 mm
  • Capacity:
    300 ml
  • Height:
    381 mm
  • Cat. No.:
    89091-314
  • Environmental Certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact