Orders LinkContactUsLinkComponent
Drop-Ceiling Leak Diversion System, New Pig
Drop-Ceiling Leak Diversion System, New Pig
61 x 121.9 cm (2 x 4')
Catalog # 89073-898
Supplier:   New Pig
Supplier #:  TLS367
Drop-Ceiling Leak Diversion System, New Pig
Catalog # 89073-898
Supplier:  New Pig
Drop-Ceiling Leak Diversion System, New Pig
Drop-Ceiling Leak Diversion System, New Pig
61 x 121.9 cm (2 x 4')
Price
$244.86
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Remove damaged tile and replace it with a leak diversion system panel to channel away liquids
Includes drain hose and clips to secure hose to ceiling grids. Flow rate of 1 gallon per minute when used with included hose. After leak has been repaired, simply uninstall and store for later use.
Optional Splash Pads for Drop-Ceiling Leak Diverters prevents annoying dripping sound of water hitting plastic. Diverter fiberbond pad is treated with an anti-microbial solution to help slow the growth of mold and mildew. Low-profile, light-weight pad will not interfere with above-ceiling structures. Each pad can be used in a large drop-ceiling diverter system or cut in half to use in two small systems.
Minimizes wet floors, damaged equipment and disturbances to workers.
Specifications
  • Dimensions:
    61 x 121.9 cm (2 x 4')