Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Protector® Demonstration Hoods, Labconco®

584 mm (23"), 914 mm (36"), 22⁸/₁₀", No, 75 lbs.
Catalog # 89000-162
Supplier:   Labconco
 :  3944300
Protector® Demonstration Hoods, Labconco®
Catalog # 89000-162
Supplier:  Labconco
Protector® Demonstration Hoods, Labconco®
Protector® Demonstration Hoods, Labconco®

584 mm (23"), 914 mm (36"), 22⁸/₁₀", No, 75 lbs.

Price
$4,900.00
Log in for availability
Each
 
Product Details & Documents

Designed for classroom use, these lightweight hoods feature an epoxy-coated aluminum frame with tempered safety glass front sash, sides, rear baffle, and top.

Hood systems include a white, epoxy-coated tubular steel cart with a black 2.5cm (1") thick solid epoxy dished work surface, a handle bar, a bottom shelf, and 10.2 cm (4") diameter toe-locking casters. Also included is a FilterMate™ Portable Exhauster, which requires either two carbon filters or one carbon filter and one HEPA filter (filters sold separately).

The organic carbon filter cell is a type AC filter that contains 5.4 kg (12 lbs.) of granular activated carbon media suitable for organic solvents including acetone, ethyl acetate, toluene, and xylene. The formaldehyde carbon filter cell is a type C1 filter that contains 6.4 kg (14 lbs.) of impregnated carbon media suitable for vapors from formaldehyde. The ammonia filter cell is a type CX filter that contains 7.3 kg (16 lbs.) of impregnated carbon media suitable for ammonia and low-molecular weight amines. The HEPA filter cell is 99.99% efficient on particulates 0.3 µm in size; however, it should not be used to capture biohazardous materials or volatile organic material. The syringe kit is for carbon saturation detection (detector tubes sold separately).

Hoods have a nominal width of 91.4 cm (3') and an interior depth of 58.4 cm (23") to easily accommodate a variety of apparatus. Hoods feature the Clean-Sweep™ air foil that allows air to sweep the work surface for maximum containment; two utility ports with iris openings for connection of tubing, cords, and cables; and an ergonomic angled sash that reduces glare. Flexible exhaust options enable the hood to be connected to ductwork for exhausting to the outside or to the optional FilterMate™ Portable Exhauster (see 82010-632 series) for filtered removal of fumes and/or particulates.

 : All hoods conform to ASHRAE 110-1995 standards. The 115 V models are ETL and ETL-C listed. The 230 V models are CE marked.

 : Hoods 89000-162 and 89000-104 require a connection to a remote blower, house exhaust, or FilterMate™ Portable Exhauster (see 82010-632 series). They also require a work surface and a supporting base. For Guardian airflow monitors, see 82007-280 series. For exhaust dampers, remote blowers, transition adapters for connecting ductwork, and other accessories, contact your VWR sales representative.

Specifications
  • Shipping weight:
    75 lbs.
  • Depth:
    584 mm (23")
  • Width:
    914 mm (36")
  • Enclosure height:
    22⁸/₁₀"
  • Rediship:
    No
Accessories

Product Information

  • Catalog #
    Rediship
    Shipping weight
    Availability
    Price
  • Solid Epoxy Dished Work Surface for 3' Models
    Solid Epoxy Dished Work Surface for 3' Models
    82010-692
    Yes
    72 lbs.
    Log in for availability
    Each - $1,272.24

Product Information

  • Catalog #
    Rediship
    Shipping weight
    Availability
    Price
  • Fluorescent Light Kit, 115V, 60Hz, 10A
    Fluorescent Light Kit, 115V, 60Hz, 10A
    82010-710
    No
    15 lbs.
    Log in for availability
    Each - $1,220.00

Product Information

  • Catalog #
    Rediship
    Shipping weight
    Availability
    Price
  • Fluorescent Light Kit, 230V, 50Hz, 10A
    Fluorescent Light Kit, 230V, 50Hz, 10A
    82010-712
    No
    15 lbs.
    Log in for availability
    Each - $1,300.00

Product Information

  • Catalog #
    Rediship
    Shipping weight
    Availability
    Price
  • Cart, White Epoxy-Coated Tubular Steel
    Cart, White Epoxy-Coated Tubular Steel
    89002-372
    No
    50 lbs.
    Log in for availability
    Each - $2,090.00

Product Information

  • Catalog #
    Shipping weight
    Availability
    Price
  • Syringe Kit
    Syringe Kit
    82010-732
    0.5 kg (1 lbs.)
    Log in for availability
    Each - $60.80

Product Information

  • Catalog #
    For use with
    Rediship
    Availability
    Price
  • FilterMate™ Carbon Filter, Ammonia
    FilterMate™ Carbon Filter, Ammonia
    82010-684
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Log in for availability
    Each - $790.00

Product Information

  • Catalog #
    For use with
    Rediship
    Availability
    Price
  • Bag-In/Bag-Out Bag for HEPA Filter
    Bag-In/Bag-Out Bag for HEPA Filter
    82010-688
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Log in for availability
    Each - $445.00

Product Information

  • Catalog #
    For use with
    Rediship
    Availability
    Price
  • FilterMate™ Carbon Filter, Organic
    FilterMate™ Carbon Filter, Organic
    82010-680
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Log in for availability
    Each - $460.00

Product Information

  • Catalog #
    For use with
    Rediship
    Availability
    Price
  • FilterMate™ HEPA Filter
    FilterMate™ HEPA Filter
    82010-686
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Log in for availability
    Each - $790.00

Product Information

  • Catalog #
    Shipping weight
    Availability
    Price
  • Ammonia Detector Tubes, Set of 3
    Ammonia Detector Tubes, Set of 3
    46000-246
    0.5 kg (1 lbs.)
    Log in for availability
    Each - $142.00

Product Information

  • Catalog #
    For use with
    Rediship
    Availability
    Price
  • FilterMate™ Carbon Filter, Formaldehyde
    FilterMate™ Carbon Filter, Formaldehyde
    82010-682
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Log in for availability
    Each - $790.00

Product Information

  • Catalog #
    Shipping weight
    Availability
    Price
  • Formaldehyde Detector Tubes, Set of 3
    Formaldehyde Detector Tubes, Set of 3
    46000-250
    0.5 kg (1 lbs.)
    Log in for availability
    Each - $174.00