Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Analyzer Sample Cups, Greiner Bio-One

Analyzer Sample Cups, Greiner Bio-One

Sample Cup for Hitachi® Analyzers, Conical/Skirted, 1.5 ml (0.05 oz.), 17×38 mm
Catalog # 82051-134
Supplier:   Greiner Bio-One
Supplier #:  729101
Analyzer Sample Cups, Greiner Bio-One
Catalog # 82051-134
Supplier:  Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One

Sample Cup for Hitachi® Analyzers, Conical/Skirted, 1.5 ml (0.05 oz.), 17×38 mm

Price
$242.00
Log in for availability
Case of 5,000
Minimum order quantity is 1
Product Details & Documents

Sample cups are well suited for a number of established analyzer systems including Centrifichem, Coulter Counter®, Gemsaec, Hitachi®, Technicon™, and Sysmex® systems.

  • Polyethylene or polystyrene construction
  • Suited for a number of established analyzer systems
  • Clear

Sample cup 82051-154 is constructed of polyethylene; all others are constructed of polystyrene.

Documents

Specifications
  • Capacity:
    1.5 ml (0.05 oz.)
  • Description:
    Sample Cup for Hitachi® Analyzers, Conical/Skirted
  • W×H:
    17×38 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.