Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Analyzer Sample Cups, Greiner Bio-One

Analyzer Sample Cups, Greiner Bio-One

Sample Cup for Coulter Counter® Analyzers, Flat, 25 ml (0.85 oz.), 32×60 mm
Catalog # 82050-936
Supplier:   Greiner Bio-One
Supplier #:  668102
Analyzer Sample Cups, Greiner Bio-One
Catalog # 82050-936
Supplier:  Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One

Sample Cup for Coulter Counter® Analyzers, Flat, 25 ml (0.85 oz.), 32×60 mm

Price
$327.00
Log in for availability
Case of 1,250
Product Details & Documents

Sample cups are well suited for a number of established analyzer systems including Centrifichem, Coulter Counter®, Gemsaec, Hitachi®, Technicon™, and Sysmex® systems.

  • Polyethylene or polystyrene construction
  • Suited for a number of established analyzer systems
  • Clear

Sample cup 82051-154 is constructed of polyethylene; all others are constructed of polystyrene.

Documents

Specifications
  • Capacity:
    25 ml (0.85 oz.)
  • Description:
    Sample Cup for Coulter Counter® Analyzers, Flat
  • W×H:
    32×60 mm