Sample Containers, Polystyrene, Greiner Bio-One
35 ml (1.2 oz.), Flat-Bottom Container, with Lamella Stopper, 56 mm, 31 mm, Non sterile
82050-326
: 214111
This collection of clear polystyrene sample containers is designed to meet general-purpose collection, storage, and transport requirements for most lab environments.
- Clear polystyrene construction
- Supplied sterile or nonsterile
- Flat or conical bottom styles
- Supplied with scrap caps
Containers are available with flat- or conical-bottom styles, and can be supplied sterile or nonsterile. Flat-bottom containers are supplied with plastic or metal screw caps, and conical-bottom containers are supplied with plastic, wadded screw caps.
- Sterility:Non sterile
- Width:31 mm
- Capacity:35 ml (1.2 oz.)
- Description:Flat-Bottom Container, with Lamella Stopper
- Height:56 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



