Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Protector® XVS Ventilation Stations, Labconco®

2 ft., 23", 22", 22¾", 29", 22¾", 24", 95 cfm at 80 fpm, 70 cfm at 60 fpm, Yes, 50 lbs.
  82010-736
 :  4862010
Protector® XVS Ventilation Stations, Labconco®
  82010-736
 :  
Protector® XVS Ventilation Stations, Labconco®
Protector® XVS Ventilation Stations, Labconco®

2 ft., 23", 22", 22¾", 29", 22¾", 24", 95 cfm at 80 fpm, 70 cfm at 60 fpm, Yes, 50 lbs.

 
 
 
 

This ventilated work area is ideal as a weighing station, light-duty fume hood, science classroom workstation, solvent cleaning work bay, or for veterinary pathology and cytology

The deep 58.4cm (23") interior easily accommodates a variety of equipment including large balances. Glacier white and gray, dry powder epoxy-coated enclosures have a sturdy aluminum frame with 0.5cm (3/16") thick tempered safety glass top and side walls. The angled and hinged sash with 0.6cm (1/4") thick tempered safety glass swings up for easy loading and cleaning. The sash opening on 57.8cm (223/4") high models is 20.3cm (8"); on 81.3cm (32") high models the opening is 25.4cm (10"). The sash pivots upward and locks to a loading height of 50.8cm (20") on 57.8cm (223/4") high models, and 74.3cm (291/4") on the 81.3cm (32") high models.

The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. Airflow openings pull air under the airfoil, creating a constant barrier of protection from contaminants. The upper containment sash foil bleeds air into the enclosure to direct contaminants away from the operator’s breathing zone. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. Additional side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment. Two utility ports allow passage of tubing and cords. Enclosures feature 5.1x25.4cm (2x10") rectangular exhaust collars on the top and bottom for connection to ductwork or portable exhauster.

Black, chemical-resistant, solid epoxy work surfaces are contoured to fit the dimensions of the enclosure and dished to contain spills. Base stands are made of durable 4.4cm (13/4") thick, tubular, glacier white epoxy-coated steel. The height of base stands is fully adjustable from 70–85cm (271/2–331/2") in 2.5cm (1") increments. Base stands come with either fixed leveling feet for use on uneven floor surfaces or four 12.7cm (5") non-marking casters.

Utility shelf kit includes three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure. Remote blowers 30153-980, 30153-982, and 82010-730 have a 1/4hp, totally enclosed, fan-cooled (TEFC) standard motor capable of overcoming external static pressure loss of 0.75in.H2O at 360cfm. Housing and wheel are made of corrosion-resistant, epoxy-coated steel. Blower inlet has an I.D. of 15.2cm (6"). Outlet dimensions are 10.8Wx18.7Lcm (41/4x73/8"). Applications involving high concentrations of solvents may require the explosion-proof blower.

Even, inward airflow protects the operator and contains hazardous or noxious fumes, vapors, and fine powders.

 : All systems conform to ASHRAE 110-1995 standards.

 : Work surface and stand are not included and must be purchased separately; contact your Avantor representative for more information. Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures must be connected to either a remote blower or a FilterMate portable exhauster (see 82010-632 series) for proper exhaustion. For Guardian airflow monitors, see 82007-280 series. Gray work surfaces are also available; contact your Avantor sales representative for more information.

 : VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.

 
  • Inside height:
    22"
  • Inside width:
    22¾"
  • Outside height:
    22¾"
  • Outside depth:
    29"
  • Outside width:
    24"
  • Shipping weight:
    50 lbs.
  • Inside depth:
    23"
  • Width:
    2 ft.
  • Exhaust volume:
    95 cfm at 80 fpm, 70 cfm at 60 fpm
  • Rediship:
    Yes
Accessories

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-712
    Fluorescent Light Kit, 230V, 50Hz, 10A
    No
    15 lbs.
       
    Each - $1,300.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 30153-980
    Remote Blower, 115/230V, 50Hz, 5.6/2.8A, 35.6 x 28 x 38.1 cm (14 x 11 x 15")
    No
    16 kg (35 lbs.)
       
    Each - $1,960.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-730
    Explosion-Proof Remote Blower, 115V, 60Hz, 4.4A
    No
    40 lbs.
       
    Each - $2,940.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 82003-596
    Adjustable Base Stand, Electric Hydraulic Lift
    115 V, 60 Hz
    4' Models
       
    Each - $4,950.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-722
    Lower Exhaust Transition Adapter, 6" O.D. Duct
    Yes
    5 lbs.
       
    Each - $202.14

 

  •  
    Description
    Electrical
    For use with
     
     
  • 82003-594
    Adjustable Base Stand, Electric Hydraulic Lift
    115 V, 60 Hz
    3' Models
       
    Each - $4,890.00

 

  •  
    Description
    Outside depth
    Outside height
     
     
  • 89000-180
    Telescoping Base Stand with Fixed Feet
    25½"
    27½ - 33½"
       
    Each - $1,210.00

 

  •  
    Description
    Shipping weight
     
     
  • 97006-412
    Telescoping Base Stand Shelf Kit
    9 lbs.
       
    Each - $169.00

 

  •  
    Description
    Inside depth
    Outside height
     
     
  • 82010-694
    Solid Epoxy Dished Work Surface
    1"
    26¾"
       
    Each - $1,530.00

 

  •  
    Description
    For use with
    Outside depth
     
     
  • 62111-176
    Adjustable Base Stand, Telescoping, with Fixed Feet
    3' Models
    29"
       
    Each - $1,280.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-692
    Solid Epoxy Dished Work Surface for 3' Models
    Yes
    72 lbs.
       
    Each - $1,272.24

 

  •  
    Description
    Outside depth
    Outside height
     
     
  • 82003-570
    Telescoping Base Stand with Casters
    29"
    27½ - 33½"
       
    Each - $1,590.00

 

  •  
    Description
    Outside depth
    Outside height
     
     
  • 82003-572
    Telescoping Base Stand with Casters
    29"
    27½ - 33½"
       
    Each - $1,680.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-710
    Fluorescent Light Kit, 115V, 60Hz, 10A
    No
    15 lbs.
       
    Each - $1,220.00

 

  •  
    Description
     
     
  • 97006-416
    Accessory Shelf, 4'
       
    Each - $276.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 97000-594
    Tissue Holder
    No
    0.5 kg (1 lb.)
       
    Each - $112.00

 

  •  
    Description
    For use with
    Outside depth
     
     
  • 62111-178
    Adjustable Base Stand, Telescoping, with Fixed Feet
    4' Models
    29"
       
    Each - $1,350.00

 

  •  
    Description
    For use with
    Interior dimensions
     
     
  • 82003-630
    Caster Kit for Hydraulic Lift Base Stands
       
    Each - $232.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-716
    Upper Exhaust Transition Adapter, 5" I.D. Hose
    No
    5 lbs.
       
    Each - $200.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 97000-592
    Bottle Holder
    No
    0.5 kg (1 lb.)
       
    Each - $78.00

 

  •  
    Description
    For use with
    Outside depth
     
     
  • 82003-582
    Adjustable Base Stand, Manual Hydraulic Lift
    3' Models
    29"
       
    Each - $3,880.00

 

  •  
    Description
    For use with
    Interior dimensions
     
     
  • 97010-022
    Seismic Support Bracket Kit, for Telescoping Base Stands with Fixed Feet
       
    Each - $405.00

 

  •  
    Description
    Ø
    Height
     
     
  • 82010-726
    Manual duct damper
    15.2 cm (6")
    35.6 cm (14")
       
    Each - $650.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-714
    Utility Shelf Kit (3 Shelves)
    No
    2.3 kg (5 lbs.)
       
    Each - $191.00

 

  •  
    Description
    Outside depth
    Outside height
     
     
  • 89000-178
    Telescoping Base Stand with Casters
    25½"
    27½ - 33½"
       
    Each - $1,510.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 82003-602
    Adjustable Base Stand, Electric Hydraulic Lift
    230 V, 50 Hz
    4' Models
       
    Each - $4,950.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 30153-982
    Remote Blower, 115V, 60Hz, 4.4A
    Yes
    35 lbs.
       
    Each - $1,760.00

 

  •  
    Description
    Electrical
    For use with
     
     
  • 82003-600
    Adjustable Base Stand, Electric Hydraulic Lift
    230 V, 50 Hz
    3' Models
       
    Each - $4,890.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-718
    Upper Exhaust Transition Adapter, 6" O.D. Duct
    Yes
    5 lbs.
       
    Each - $202.14

 

  •  
    Description
     
     
  • 97006-414
    Accessory Shelf, 3'
       
    Each - $203.00

 

  •  
    Description
    For use with
    Outside depth
     
     
  • 82003-584
    Adjustable Base Stand, Manual Hydraulic Lift
    4' Models
    29"
       
    Each - $3,940.00

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-724
    Exhaust Damper, Powder-Coated Steel,  15.2 cm (6") I.D.
    No
    0.5 (1)
       
    Each - $58.70

 

  •  
    Description
    Rediship
    Shipping weight
     
     
  • 82010-720
    Lower Exhaust Transition Adapter, 5" I.D. Hose
    No
    5 lbs.
       
    Each - $200.00