Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER TRANSFR LNGLIF 20X500ML
Catalog # 81002-126
Supplier:
NuSep
Expand image
BUFFER TRANSFR LNGLIF 20X500ML
Catalog # 81002-126
Supplier:
NuSep
Supplier Number:
BG-168