Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Low Density Flat Poly Liners, Associated Bag

Yellow, Step-on cans (liner circumference 36"), 18×24", 4 mil.
  80094-530
 :  71-6-30Y
Low Density Flat Poly Liners, Associated Bag
  80094-530
Low Density Flat Poly Liners, Associated Bag
Low Density Flat Poly Liners, Associated Bag

Yellow, Step-on cans (liner circumference 36"), 18×24", 4 mil.

 
$429.60
   
Case of 250
 

Sort and identify different types of waste.

Strong, bottom-sealed liners are free of heavy metals, making them safe for landfill or incineration.

 
  • Color:
    Yellow
  • Thickness:
    4 mil.
  • W×L:
    18×24"
  • Fits:
    Step-on cans (liner circumference 36")