Orders LinkContactUsLinkComponent
Shell Vials
Shell Vials
Vials, 1.00 ml, Flat bottom, 8×40 mm, Glass, Clear, Yes, SepCap™, Natural
Catalog # 66011-212
Shell Vials
Catalog # 66011-212
Supplier:  
Shell Vials
Shell Vials
Vials, 1.00 ml, Flat bottom, 8×40 mm, Glass, Clear, Yes, SepCap™, Natural
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
The 1 ml, 8×35 mm vials are designed for use in Alcott model 728 autosamplers. The 1 ml, 8×40 mm vials are specifically designed for Waters® 96-position trays. The 2 ml vial will work on any standard size 2 ml tray. The 4 ml vial is compatible with Waters® 48-position trays. Shell vials are manufactured from borosilicate glass unless otherwise specified.
  • Molded sealing ring secures sample containment
SepCap™ closures are designed for easy syringe needle penetration and easy push-in/pull-out from vial.
Conical-style Polyspring® inserts are available in borosilicate glass or polypropylene.
Certifications: Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.
Delivery information: Vials are supplied with polyethylene push-in caps or polypropylene SepCap™ closures. Convenience kits include vials and caps with pre-inserted septa.
Specifications
  • Cap included:
    Yes
  • Cap type:
    SepCap™
  • Capacity:
    1.00 ml
  • Color:
    Clear
  • Description:
    Vials
  • Material:
    Glass
  • Diameter ×H:
    8×40 mm
  • Cap color:
    Natural
  • Bottom style:
    Flat bottom