Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pliers, Replaceable Tip, Straight, Excelta

Pliers, Replaceable Tip, Straight, Excelta

Pliers, replaceable tip, 5 star quality, Acetal, 143.75 mm (5.75"), 3.75×3.75 mm (0.15×0.15"), Straight
Catalog # 63042-552
Supplier:   Excelta
Supplier #:  531A-US
Pliers, Replaceable Tip, Straight, Excelta
Catalog # 63042-552
Supplier:  Excelta
Pliers, Replaceable Tip, Straight, Excelta
Pliers, Replaceable Tip, Straight, Excelta

Pliers, replaceable tip, 5 star quality, Acetal, 143.75 mm (5.75"), 3.75×3.75 mm (0.15×0.15"), Straight

Price
$318.74
Log in for availability
Each
Product Details & Documents

These pliers are useful in straightening leads and wires when metal-to-metal contact is not desired.

  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use

This series of pliers offer soft jaw inserts to protect delicate parts. The jaw inserts are factory replaceable, and offered in Acetal, Brass and Nylon. The overall length is 5.75". They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs.

Specifications
  • Description:
    Pliers, replaceable tip, 5 star quality
  • Length:
    143.75 mm (5.75")
  • Material:
    Acetal
  • Curvature:
    Straight
  • Tip size:
    3.75×3.75 mm (0.15×0.15")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.