Orders LinkContactUsLinkComponent
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
250 ml, 38-430
Catalog # 62401-628
Supplier #:  355394
Trypsinizing Flasks, with Cap, Wheaton
Catalog # 62401-628
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
250 ml, 38-430
Price
$90.50
Log in for availability
Case of 1
Minimum order quantity is 1
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
  • Cap Size:
    38-430
  • Capacity:
    250 ml