Orders LinkContactUsLinkComponent
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
500 ml
Catalog # 62401-560
Supplier #:  354242
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Catalog # 62401-560
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
500 ml
Price
$77.70
Log in for availability
Case of 1
Minimum order quantity is 1
Product Details & Documents
Designed for use on either rotary or reciprocating shakers.
  • Autoclavable
  • Specially designed to maximize aeration for improved growth
  • Pinched neck design to reduce splashing on flasks with four baffles
Feature a marking spot on the side of the flask and a break-resistant lip, which accommodates a cotton plug or stopper. Made of wheaton 33® low-extractable borosilicate glass, which meets ASTM Type I Class A and USP Type I standards.
Specifications
  • Capacity:
    500 ml