Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dounce Tissue Grinders, Wheaton

Dounce Tissue Grinders, Wheaton

15 ml, 15O.D. x 82L mm
Catalog # 62400-642
Supplier #:  357544
Dounce Tissue Grinders, Wheaton
Catalog # 62400-642
Dounce Tissue Grinders, Wheaton
Dounce Tissue Grinders, Wheaton

15 ml, 15O.D. x 82L mm

Price
$499.00
Log in for availability
Case of 2
Product Details & Documents

Designed for fine particle size reduction without damage to cell nuclei

Made of low extractable borosilicate glass. Supplied with 2 pestles. The large clearance pestle is used for initial sample reduction; the narrow clearance pestle forms the final homogenate. Mortar has large reservoir and pouring lip.

For small cells such as E. coli, proteins, serum, and malignant tissues.

Loose pestle clearance: 0.0889 - 0.127 mm (0.0035 - 0.0050")

Tight pestle clearance: 0.0254 - 0.0762 mm (0.0010 - 0.0030")

Specifications
  • Capacity:
    15 ml
  • Chamber dimensions:
    15O.D. x 82L mm