Orders LinkContactUsLinkComponent
Five-Channel Timer, Sper Scientific
Five-Channel Timer, Sper Scientific
5 channel timer, certified, 2⅞×2½×½", 2 oz.
Catalog # 62364-186
Supplier:   Sper Scientific
Supplier #:  810015C
Five-Channel Timer, Sper Scientific
Catalog # 62364-186
Supplier:  Sper Scientific
Five-Channel Timer, Sper Scientific
Five-Channel Timer, Sper Scientific
5 channel timer, certified, 2⅞×2½×½", 2 oz.
Price
$179.34
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
This digital count down/count up timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours, plus clock button.
  • Extra large display indicates channels in use
  • Each one has a distinctive 1 minute alarm
Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and magnet.
Certifications: N.I.S.T. traceable certificate of compliance for the certified option.
Specifications
  • Description:
    5 channel timer, certified
  • W×H×D:
    2⅞×2½×½"
  • Weight:
    2 oz.