Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®

Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®

7-Day Chart, 0...185 °F/°C
Catalog # 61280-321
Supplier:   Dickson
Supplier #:  C665
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Catalog # 61280-321
Supplier:  Dickson
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®

7-Day Chart, 0...185 °F/°C

Price
$121.70
Log in for availability
Pack of 60
Minimum order quantity is 1
Product Details & Documents

Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, and user-selectable recording times and temperature ranges

Increased shielding eliminates electronic noise interference and allows smoother pen movement.

Ordering information: Recorders include one red pen, one blue pen, AC adapter, and 9V battery. Charts must be purchased separately.

Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

Specifications
  • Temperature range:
    0...185 °F/°C
  • Description:
    7-Day Chart
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • NIST Traceable Calibration 1-Point Service
    NIST Traceable Calibration 1-Point Service
    13450-754
    Log in for availability
    Each - $184.34

Product Information

  • Catalog #
    Availability
    Price
  • NIST & A2LA Traceable Calibration 3-Point Service
    NIST & A2LA Traceable Calibration 3-Point Service
    13450-756
    Log in for availability
    Each - $537.04

Product Information

  • Catalog #
    Availability
    Price
  • Pens, Three Red and Three Blue
    Pens, Three Red and Three Blue
    61280-134
    Log in for availability
    Pack of 6 - $154.52

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.