Orders LinkContactUsLinkComponent
pH Test Papers, Precision Laboratories
pH Test Papers, Precision Laboratories
Litmus Blue, 8 Books of 25 Strips
Catalog # 60788-001
Supplier #:  380-1V-8-25
pH Test Papers, Precision Laboratories
Catalog # 60788-001
pH Test Papers, Precision Laboratories
pH Test Papers, Precision Laboratories
Litmus Blue, 8 Books of 25 Strips
Price
$34.22
Log in for availability
Pack of 200
Product Details & Documents
Available in 8 books of 25 sheets, or vials containing 100 sheets.
Specifications
  • Description:
    Litmus Blue
  • Size:
    8 Books of 25 Strips