Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

50 L (13 gal.), 61 x 30 x 30 cm (24 x 12 x 12")
  60463-113
Supplier #:  14200-0020
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
  60463-113
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

50 L (13 gal.), 61 x 30 x 30 cm (24 x 12 x 12")

 
$996.00
   
Each
 

Tanks have stepped rim for strength and overlapping skirted cover

Polypropylene tank is molded in one piece and is autoclavable

Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.

Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.

Documents

 
  • Volume:
    50 L (13 gal.)
  • L×W×D:
    61 x 30 x 30 cm (24 x 12 x 12")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.