Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Synthware Sure-Grip Separatory Funnels

With glass stopper, 2000 ml, 4 mm, 27
  60004-254
 :  F68272LA
Synthware Sure-Grip Separatory Funnels
  60004-254
Synthware Sure-Grip Separatory Funnels
Synthware Sure-Grip Separatory Funnels

With glass stopper, 2000 ml, 4 mm, 27

 
$318.12
   
Each
 
 

Globe-shaped separatory funnel features a long stopper neck and a straight section between the funnel body and stopcock that provides a secure grip while shaking

  • Sure-grip design
  • Heavy wall borosilicate construction
  • Ground glass joint

The straight section also aids in clamping. Available with PTFE stopcocks. Funnels are supplied complete with one PTFE or hollow glass stopper.

 
  • Capacity:
    2000 ml
  • Description:
    With glass stopper
  • Stopper no.:
    27
  • Stopcock size:
    4 mm