Synthware Round-Bottom Flasks with Three Necks, Angled
250 ml, 19/22, 14/20
Catalog # 60002-720
Supplier: Kemtech America
: F430250
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-720
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
- Capacity:250 ml
- Center neck [ST] joint:19/22
- Side neck ST joints:14/20