Synthware Round-Bottom Flasks with Three Necks, Angled
100 ml, 14/20, 14/20
60002-706
Supplier #: F431100
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
- Capacity:100 ml
- Center neck [ST] joint:14/20
- Side neck ST joints:14/20