Synthware Round-Bottom Flasks with Three Necks, Angled
50 ml, 24/40, 24/40
Catalog # 60002-704
Supplier: Kemtech America
Supplier #: F434450
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-704
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
- Capacity:50 ml
- Center neck [ST] joint:24/40
- Side neck ST joints:24/40