Synthware Extraction Thimbles, Glass
Large extractor size, 45×130 mm, Coarse
60002-222
Supplier #: E114513C
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
- Porosity:Coarse
- O.D.×H:45×130 mm
- Fits:Large extractor size