Angled Shelf Carts, Eagle MHC
60", 18", 72", Chrome
58202-564
: M1860C-4
Features four wire angled shelves which adjust on 1' vertical increments and the slope of each shelf, four posts, and four 5" resilient swivel casters, two with brake
Shelves feature 3-1/4" upturn at front and 1-1/4" downturn at rear.
- Width:18"
- Height:72"
- Length:60"
- Finish:Chrome



