Orders LinkContactUsLinkComponent
Full Size Security Units, Eagle MHC
Full Size Security Units, Eagle MHC
Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes, 51¼", 27¼", 69", 83.9 kg (185 lbs.)
Catalog # 58202-382
Supplier:   Eagle Group
Supplier #:  CSC2448
Full Size Security Units, Eagle MHC
Catalog # 58202-382
Supplier:  Eagle Group
Full Size Security Units, Eagle MHC
Full Size Security Units, Eagle MHC
Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes, 51¼", 27¼", 69", 83.9 kg (185 lbs.)
Price
$3,678.92
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Security units are ideal for safely storing and transporting costly materials and items subject to pilferage.
  • Heavy-gauge open-wire construction keeps contents visible for quick inventory checks
  • Variety of sizes are available in stationary models or with casters for easy mobility
  • All units are plated with a durable easy-to-clean finish
  • Quick-action locking feature with hasps for padlock
  • All units feature full-access double door swinging out at approximately 270°
Mobile security units feature 5" (127 mm) diameter swivel stem casters with poly tread, two with brake. Casters on stainless steel units are stainless steel. Casters on chrome and EAGLEgard® units are nickel-plated.
Packaging: Shipped knocked down for easy assembly and reduced transportation costs.
Specifications
  • Width:
    27¼"
  • Description:
    Chrome Mobile, Full-Size Security Unit. Includes Four Poly Stem Swivel Casters 5×11/4", Two With Brakes
  • Height:
    69"
  • Length:
    51¼"
  • Weight:
    83.9 kg (185 lbs.)
Accessories
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-004
    36L×14" W
    3.6 kg (8 lbs.)
    Log in for availability
    Each - $143.63
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-006
    48L×14" W
    4.5 kg (10 lbs.)
    Log in for availability
    Each - $163.29
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-024
    30L×24" W
    5.0 kg (11 lbs.)
    Log in for availability
    Each - $183.09
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-026
    36L×24" W
    5.9 kg (13 lbs.)
    Log in for availability
    Each - $188.50
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-030
    48L×24" W
    7.3 kg (16 lbs.)
    Log in for availability
    Each - $224.42
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-036
    36L×30" W
    7.7 kg (17 lbs.)
    Log in for availability
    Each - $276.44
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-040
    60L×30" W
    11.4 kg (25 lbs.)
    Log in for availability
    Each - $350.08
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-038
    48L×30" W
    9.1 kg (20 lbs.)
    Log in for availability
    Each - $316.03
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-068
    48L×24" W
    7.3 kg (16 lbs.)
    Log in for availability
    Each - $858.17
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-070
    60L×24" W
    9.5 kg (21 lbs.)
    Log in for availability
    Each - $1,046.57
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Eaglegard® Hybrid Epoxy with Microgard®, Wire Shelf
    Eaglegard® Hybrid Epoxy with Microgard®, Wire Shelf
    58202-082
    48L×24" W
    7.3 kg (16 lbs.)
    Log in for availability
    Each - $276.44
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-002
    30L×14" W
    3.2 kg (7 lbs.)
    Log in for availability
    Each - $134.69
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-920
    30L×14" W
    3.2 kg (7 lbs.)
    Log in for availability
    Each - $424.43
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-924
    60L×14" W
    6.4 kg (14 lbs.)
    Log in for availability
    Each - $172.98
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-934
    30L×24" W
    5.0 kg (11 lbs.)
    Log in for availability
    Each - $647.33
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-936
    36L×30" W
    7.7 kg (17 lbs.)
    Log in for availability
    Each - $868.46
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-938
    48L×30" W
    9.1 kg (20 lbs.)
    Log in for availability
    Each - $1,012.92
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Stainless Steel Wire Shelf
    Stainless Steel Wire Shelf
    58202-940
    60L×30" W
    11.4 kg (25 lbs.)
    Log in for availability
    Each - $1,123.48
Product Information
  • Catalog #
    Dimensions
    Weight
    Availability
    Price
  • Chrome Wire Shelf
    Chrome Wire Shelf
    58202-032
    60L×24" W
    9.5 kg (21 lbs.)
    Log in for availability
    Each - $267.50