Orders LinkContactUsLinkComponent
Drum Tray, Eagle Manufacturing
Drum Tray
  56617-343
 :  1614
Drum Tray, Eagle Manufacturing
  56617-343
Drum Tray, Eagle Manufacturing
Drum Tray, Eagle Manufacturing
Drum Tray
$335.33
Each
Tray with reservoir keeps container away from spills
Ideal for use under drums during dispensing/filling operations. Holds up to 34L (9gal.) of spills when used with a 208.2L (55gal.) drum. Constructed of high-density polyethylene. Use drum tray dolly to easily transport drums between work areas without compromising spill protection.
  • Description:
    Drum Tray