Double Drum Secondary Containment Workstation, Eagle Manufacturing
Double Drum Work Station, 130 lbs.
Catalog # 56617-329
Supplier: Eagle Manufacturing
Supplier #: 1626
Double Drum Secondary Containment Workstation, Eagle Manufacturing
Catalog # 56617-329
Supplier: Eagle Manufacturing
Jump to:
Product Details & Documents
For secure indoor or outdoor drum storage.
Heavy-duty polyethylene unit in safety yellow holds two 208.2L (55gal.) drums. Use as a collection station or dispensing center for most hazardous chemicals. Secondary spill capacity of 454L (120gal.). Forkliftable. Comes complete with locks.
Meets EPA requirement 40 CFR 264.175.
Specifications
- Description:Double Drum Work Station
- Weight:130 lbs.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



