Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Generators for Polytron® PT Models, Kinematica

Generators for Polytron® PT Models, Kinematica

PT-DA 12 / 2EC-E123, Saw Tooth, 2.00 - 250 ml, 12 mm, 12×123 mm, PT 1200 E, PT 1300 D, PT 2500 E
Catalog # 53513-440
Supplier:   Kinematica
Supplier #:  11030062
Generators for Polytron® PT Models, Kinematica
Catalog # 53513-440
Supplier:  Kinematica
Generators for Polytron® PT Models, Kinematica
Generators for Polytron® PT Models, Kinematica

PT-DA 12 / 2EC-E123, Saw Tooth, 2.00 - 250 ml, 12 mm, 12×123 mm, PT 1200 E, PT 1300 D, PT 2500 E

Price
$3,214.12
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications
  • Ø:
    12 mm
  • Description:
    PT-DA 12 / 2EC-E123, Saw Tooth
  • Diameter ×L:
    12×123 mm
  • For use with:
    PT 1200 E, PT 1300 D, PT 2500 E
  • Sample volume:
    2.00 - 250 ml